5-Chloro-4-nitro-1H-imidazole 95%

Supplier: Ambeed
Warning

A785971-100MG A785971-10G A785971-250MG
103840-994EA 173 USD
103840-994 76820-486 76820-488 76820-490
5-Chloro-4-nitro-1H-imidazole 95%
5-Chloro-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₂ClN₃O₂
MW: 147.52 g/mol
Boiling Pt: 427.2 °C (760 mm Hg)
Flash Pt: 212.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06240217
CAS Number: 57531-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR