5-Chloro-3-iodopyridin-2-ol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-iodopyridin-2(1H)-one

A275593-250MG A275593-1G A275593-10G
103813-268EA 25 USD
103813-268 103853-192 77014-508 77093-046 77612-598
5-Chloro-3-iodopyridin-2-ol 95%
5-Chloro-3-iodopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃ClINO
MW: 255.44 g/mol
Boiling Pt: 295.6±40.0 °C (760 mm Hg)
Flash Pt: 132.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08276120
CAS Number: 188057-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR