5-Chloro-3-hydroxypyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-pyridinol, 3-Chloro-5-hydroxypyridine, 5-Chloro-3-pyridinol
A168322-100G
A168322-10G
A168322-500G
A168322-1G
A168322-25G
A168322-5G
103806-276EA
5
USD
103806-276
103844-532
76872-288
76872-290
76872-292
77081-588
77081-590
77081-592
5-Chloro-3-hydroxypyridine 97%
5-Chloro-3-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₅H₄ClNO Melting Pt: 160…162 °C Flash Pt: 150.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006379 CAS Number: 74115-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...