5-Chloro-3H-imidazo[4,5-b]pyridine 97%

Supplier: Ambeed
Warning

A181594-100MG A181594-250MG A181594-1G A181594-10G
103807-624EA 22 USD
103807-624 103846-000 76784-594 76784-596 77083-534 77593-160
5-Chloro-3H-imidazo[4,5-b]pyridine 97%
5-Chloro-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C6H4ClN3
MW: 153.5700 g/mol
Flash Pt: 237.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646134
CAS Number: 52090-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR