5-Chloro-3-fluoro-2-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A163097-100MG
A163097-250MG
A163097-1G
A163097-5G
103805-686EA
123
USD
103805-686
103843-944
76872-102
76872-104
77320-028
77320-030
5-Chloro-3-fluoro-2-nitropyridine 98%
5-Chloro-3-fluoro-2-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₂ClFN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD19690192 CAS Number: 1064783-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...