5-Chloro-1-tetralone 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3,4-dihydronaphthalen-1(2H)-one

A104491-100MG A104491-250MG A104491-1G
103796-722EA 36 USD
103796-722 103822-436 76765-394 76765-396 77309-302
5-Chloro-1-tetralone 95%
5-Chloro-1-tetralone

Precaución: for Research Use Only

Formula: C₁₀H₉ClO
MW: 180.631 g/mol
Boiling Pt: 306.2 °C (760 mm Hg)
Melting Pt: 65.5…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08544416
CAS Number: 26673-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR