5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%
Supplier: Ambeed
|
Warning
|
A252543-100MG
A252543-250MG
A252543-1G
A252543-5G
103812-050EA
60
USD
103812-050
103851-894
76826-998
76827-000
77091-310
77091-312
5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%
5-Chloro-3-(trifluoromethyl)pyridin-2-ol
Precaución: for Research Use Only
Formula:
C₆H₃ClF₃NO Boiling Pt: 213 °C (760 mm Hg) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD20275098 CAS Number: 1214342-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...