5-Chloro-3-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: Ambeed
Warning

A233804-100MG A233804-250MG
103810-908EA 238 USD
103810-908 77890-355 77890-356
5-Chloro-3-(trifluoromethyl)-1H-pyrazole ≥95%
5-Chloro-3-(trifluoromethyl)-1H-pyrazole

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₂ClF₃N₂
Storage Temperature: Refrigerator
MDL Number: MFCD07375186
CAS Number: 131797-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR