5-Chloro-2-pyridineboronic acid 95% mix 50% LiOH mix 50% LiOH
Supplier: Ambeed
|
Warning
|
A174866-1G
A174866-5G
A174866-10G
A174866-250MG
A174866-25G
103806-982EA
38
USD
103806-982
103845-282
77082-590
77082-592
77273-832
77273-834
77301-356
5-Chloro-2-pyridineboronic acid 95% mix 50% LiOH mix 50% LiOH
5-Chloro-2-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₅H₅BClNO₂ Boiling Pt: 317.7 °C (760 mm Hg) Flash Pt: 145.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD07368862 CAS Number: 652148-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...