5-Chloro-2-nitropyridine 97%

Supplier: Ambeed
Warning

A345583-25G A345583-5G A345583-100G A345583-10G A345583-1G A345583-500G
103836-912EA 6 USD
103836-912 76839-978 76839-980 76989-516 76989-518 76989-520 76989-522
5-Chloro-2-nitropyridine 97%
5-Chloro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
Boiling Pt: 292.3 °C (760 mm Hg)
Melting Pt: 148…150 °C
Flash Pt: 120.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03092916
CAS Number: 52092-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR