5-Chloro-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A125559-100G A125559-10G A125559-25G A125559-1G A125559-5G
103800-694EA 11 USD
103800-694 77271-760 77271-762 77271-764 77298-062 77312-842
5-Chloro-2-nitrobenzaldehyde 98%
5-Chloro-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClNO₃
Boiling Pt: 310.2 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 141.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007289
CAS Number: 6628-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR