5-Chloro-2-nitroanisole 98%
Supplier: Ambeed
|
Warning
|
A127190-100G
A127190-10G
A127190-1G
A127190-25G
A127190-5G
103800-930EA
8
USD
103800-930
77136-778
77136-780
77298-190
77313-174
77313-176
5-Chloro-2-nitroanisole 98%
5-Chloro-2-nitroanisole
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₃ Boiling Pt: 289.1 °C (760 mm Hg) Melting Pt: 70…72 °C Flash Pt: 128.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007288 CAS Number: 6627-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...