5-Chloro-2-methylindole 98%

Supplier: Ambeed
Warning

A108365-10G A108365-250MG A108365-25G A108365-1G A108365-5G
103797-336EA 10 USD
103797-336 103823-116 77257-534 77257-536 77257-538 77283-712 77309-962
5-Chloro-2-methylindole 98%
5-Chloro-2-methylindole

Precaución: for Research Use Only

Formula: C₉H₈ClN
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00005619
CAS Number: 1075-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR