5-Chloro-2-methyl-3-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-nitro-o-toluic acid, 3-Carboxy-5-chloro-2-methylnitrobenzene

A433935-1G A433935-250MG
103827-568EA 15 USD
103827-568 103838-636 76842-350 77016-614
5-Chloro-2-methyl-3-nitrobenzoic acid 95%
5-Chloro-2-methyl-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
Boiling Pt: 352.1±42.0 °C (760 mm Hg)
Flash Pt: 166.7 °C
Storage Temperature: Ambient
MDL Number: MFCD09036377
CAS Number: 154257-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR