5-Chloro-2-methyl-2H-indazole 95%

Supplier: Ambeed
Warning

A100562-100MG A100562-250MG A100562-1G
103795-512EA 97 USD
103795-512 76780-380 76780-382 77296-758
5-Chloro-2-methyl-2H-indazole 95%
5-Chloro-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
Boiling Pt: 299.8 °C (760 mm Hg)
Flash Pt: 135.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11977537
CAS Number: 541539-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR