5-Chloro-2-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A129471-250MG A129471-1G
103801-284EA 22 USD
103801-284 76767-118 77298-342
5-Chloro-2-methoxypyrimidine 98%
5-Chloro-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.559 g/mol
Boiling Pt: 234.5 °C (760 mm Hg)
Melting Pt: 50…55 °C
Flash Pt: 97.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07440091
CAS Number: 38373-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR