5-Chloro-2-methoxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A258577-1G
A258577-25G
A258577-5G
103812-388EA
6
USD
103812-388
103852-264
77091-798
77091-800
77091-802
5-Chloro-2-methoxybenzaldehyde 97%
5-Chloro-2-methoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂ Melting Pt: 70…74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00033854 CAS Number: 7035-09-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...