5-Chloro-2-iodophenol 98%

Supplier: Ambeed
Warning

A125634-250MG A125634-5G A125634-10G
103800-708EA 20 USD
103800-708 76766-874 77312-854 77624-046
5-Chloro-2-iodophenol 98%
5-Chloro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
Boiling Pt: 218.9 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09753473
CAS Number: 136808-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR