5-Chloro-2-iodoaniline 98%
Supplier: Ambeed
|
Danger
|
A579712-5G
A579712-1G
103828-858EA
6
USD
103828-858
103840-006
76844-140
77307-866
5-Chloro-2-iodoaniline 98%
5-Chloro-2-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₅ClIN MW: 253.47 g/mol Boiling Pt: 295.8 °C (760 mm Hg) Melting Pt: 40…42 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06738960 CAS Number: 6828-35-9 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...