5-Chloro-2-hydroxy-3-nitropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-nitropyridin-2-ol

A208914-10G A208914-5G
103817-898EA 10 USD
103817-898 103848-586 76986-644 77086-918
5-Chloro-2-hydroxy-3-nitropyridine 95%
5-Chloro-2-hydroxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
Boiling Pt: 201.1 °C (760 mm Hg)
Melting Pt: 64…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00114884
CAS Number: 21427-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR