5-Chloro-2-formylphenylboronic acid 95%

Supplier: Ambeed
Warning

A102481-250MG A102481-1G A102481-5G
103796-376EA 9 USD
103796-376 103822-070 77148-296 77308-952 77308-954
5-Chloro-2-formylphenylboronic acid 95%

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MW: 184.3900 g/mol
Boiling Pt: 393.8 °C (760 mm Hg)
Flash Pt: 192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425955
CAS Number: 870238-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR