5-Chloro-2-formylbenzonitrile 98%

Supplier: Ambeed
Warning

A128816-10G A128816-250MG A128816-1G
103801-186EA 80 USD
103801-186 76870-638 76870-640 77313-540
5-Chloro-2-formylbenzonitrile 98%
5-Chloro-2-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MW: 165.576 g/mol
Melting Pt: 105…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699538
CAS Number: 77532-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR