5-Chloro-2-formylbenzoic acid 97%

Supplier: Ambeed
Warning

A202179-250MG A202179-1G A202179-5G
103817-362EA 46 USD
103817-362 103847-998 76825-598 77302-794 77302-796
5-Chloro-2-formylbenzoic acid 97%
5-Chloro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
Boiling Pt: 276 °C (760 mm Hg)
Flash Pt: 117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739601
CAS Number: 4506-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR