5-Chloro-2-fluoropyridine 97%

Supplier: Ambeed
Danger

A128195-10G A128195-1G A128195-25G A128195-5G
103801-086EA 5 USD
103801-086 76983-538 77313-374 77313-376 77313-378
5-Chloro-2-fluoropyridine 97%
5-Chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
Flash Pt: 52.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112513
CAS Number: 1480-65-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR