5-Chloro-2-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A174117-100G
A174117-10G
A174117-500G
A174117-25G
A174117-5G
103845-218EA
6
USD
103845-218
76769-316
76769-318
76769-320
77082-506
77301-318
5-Chloro-2-fluorobenzoic acid 98%
5-Chloro-2-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄ClFO₂ Boiling Pt: 274.7 °C (760 mm Hg) Melting Pt: 152…153 °C Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00665762 CAS Number: 394-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...