5-Chloro-2-fluorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A226311-100G
A226311-10G
A226311-1G
A226311-500G
A226311-25G
A226311-5G
103850-138EA
9
USD
103850-138
76770-990
76770-992
76770-994
76770-996
77088-864
77088-866
5-Chloro-2-fluorobenzaldehyde 98%
5-Chloro-2-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄ClFO Melting Pt: 34…39 °C Flash Pt: 95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094518 CAS Number: 96515-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...