5-Chloro-2-ethylaniline 97%

Supplier: Ambeed
Warning

A167932-1G A167932-100MG A167932-250MG A167932-25G
103806-230EA 128 USD
103806-230 103844-488 77081-522 77191-324 77191-326 77191-328
5-Chloro-2-ethylaniline 97%
5-Chloro-2-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 252.6 °C (760 mm Hg)
Flash Pt: 106.6 °C
Storage Temperature: Ambient
MDL Number: MFCD20694404
CAS Number: 3843-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR