5-Chloro-2-ethoxy-nicotinic acid 97%
Supplier: Ambeed
|
Warning
|
A150346-100MG
A150346-250MG
A150346-1G
A150346-5G
103804-148EA
31
USD
103804-148
103842-284
76997-314
76997-316
77317-798
77317-800
5-Chloro-2-ethoxy-nicotinic acid 97%
5-Chloro-2-ethoxy-nicotinic acid
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₃ Boiling Pt: 313.3±37.0 °C (760 mm Hg) Melting Pt: 114…116 °C Flash Pt: 143.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD07403147 CAS Number: 68359-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...