5-Chloro-2-cyano-3-nitropyridine 98%
Supplier: Ambeed
|
Danger
|
A250015-1G
A250015-25G
A250015-5G
A250015-100G
A250015-10G
103811-908EA
11
USD
103811-908
103851-748
77091-096
77091-098
77091-100
77276-304
77276-306
5-Chloro-2-cyano-3-nitropyridine 98%
5-Chloro-2-cyano-3-nitropyridine
Formula:
C6H2ClN3O2 Boiling Pt: 329.5 °C (760 mm Hg) Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD06657552 CAS Number: 181123-11-5 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...