5-Chloro-2,4-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A308393-1G A308393-25G A308393-5G A308393-100G A308393-500G
103814-558EA 5 USD
103814-558 103835-734 76838-490 76838-492 76838-494 77114-564 77114-566
5-Chloro-2,4-difluorobenzoic acid 97%
5-Chloro-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C7H3ClF2O2
Boiling Pt: 272.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06658163
CAS Number: 130025-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR