5-Chloro-2,3-dimethylpyrazine 98%

Supplier: Ambeed
Warning

A115531-100MG A115531-250MG A115531-5G
103798-582EA 61 USD
103798-582 103824-440 76822-078 76822-080 77297-378
5-Chloro-2,3-dimethylpyrazine 98%
5-Chloro-2,3-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.586 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189517
CAS Number: 59489-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR