5-Chloro-2-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A141980-10G A141980-1G A141980-25G A141980-5G
103803-088EA 16 USD
103803-088 76767-808 77316-290 77316-292 77316-294
5-Chloro-2-(trifluoromethyl)benzaldehyde 98%
5-Chloro-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
Boiling Pt: 225.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03412204
CAS Number: 90381-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR