5-Chloro-1-methylimidazole 97%

Supplier: Ambeed
Warning

A192738-5G A192738-10G A192738-1G A192738-25G
103816-476EA 18 USD
103816-476 103847-086 77084-962 77287-504 77302-316 77302-318
5-Chloro-1-methylimidazole 97%
5-Chloro-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
Boiling Pt: 82…85 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00014505
CAS Number: 872-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR