5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A111028-250MG A111028-1G A111028-5G A111028-100MG
103797-778EA 58 USD
103797-778 103823-584 76982-728 77310-394 77310-396 77885-022
5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%
5-Chloro-1-methyl-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₄ClN₃O₂
Boiling Pt: 273 °C (760 mm Hg)
Melting Pt: 135…137 °C
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD21607270
CAS Number: 42098-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR