5-Chloro-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Warning

A278154-250MG A278154-5G
103813-400EA 23 USD
103813-400 103853-338 76772-268 77093-252
5-Chloro-1H-pyrrolo[3,2-b]pyridine 97%
5-Chloro-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062553
CAS Number: 65156-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR