5-Chloro-1H-pyrrolo[2,3-c]pyridine 98%

Supplier: Ambeed
Warning

A215871-100G A215871-10G A215871-25G A215871-1G A215871-5G
103818-522EA 11 USD
103818-522 103849-230 77030-672 77030-674 77030-676 77087-788 77087-790
5-Chloro-1H-pyrrolo[2,3-c]pyridine 98%
5-Chloro-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
Boiling Pt: 335.8 °C (760 mm Hg)
Flash Pt: 187.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09907859
CAS Number: 131084-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR