5-Chloro-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-7-azaindole

A323360-10G A323360-500G A323360-100G A323360-1G A323360-25G A323360-5G
103825-334EA 5 USD
103825-334 103836-270 77154-564 77154-566 77167-340 77306-288 77306-290 77306-292
5-Chloro-1H-pyrrolo[2,3-b]pyridine 98%
5-Chloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 161…162 °C
Flash Pt: 157.1 °C
Storage Temperature: Ambient
MDL Number: MFCD06659676
CAS Number: 866546-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR