5-Chloro-1H-pyrazolo[4,3-b]pyridine 97%

Supplier: Ambeed
Warning

A198997-1G A198997-25G A198997-5G A198997-10G A198997-250MG
103817-066EA 8 USD
103817-066 103847-690 77085-720 77085-722 77085-724 77152-086 77152-088
5-Chloro-1H-pyrazolo[4,3-b]pyridine 97%
5-Chloro-1H-pyrazolo[4,3-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
Boiling Pt: 323.8 °C (760 mm Hg)
Melting Pt: 201…202 °C at N/A mmHg
Flash Pt: 179.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05663984
CAS Number: 94220-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR