5-Chloro-1H-pyrazolo[3,4-c]pyridine 97%

Supplier: Ambeed
Warning

A123175-100MG A123175-10G A123175-250MG A123175-25G A123175-1G A123175-5G
103800-356EA 22 USD
103800-356 77161-454 77161-456 77161-458 77161-460 77312-472 77312-474
5-Chloro-1H-pyrazolo[3,4-c]pyridine 97%
5-Chloro-1H-pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05663983
CAS Number: 76006-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR