5-Chloro-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A231510-250MG A231510-1G A231510-5G
103810-778EA 72 USD
103810-778 103850-528 76771-170 77303-914 77303-916
5-Chloro-1H-pyrazole-3-carboxylic acid 97%
5-Chloro-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O₂
Boiling Pt: 445.6 °C (760 mm Hg)
Melting Pt: 268…271 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09608056
CAS Number: 881668-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR