5-Chloro-1H-indole-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A112452-100MG A112452-250MG A112452-5G
103798-016EA 103 USD
103798-016 103823-852 77008-980 77008-982 77310-658
5-Chloro-1H-indole-2-carbaldehyde 97%
5-Chloro-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆ClNO
Boiling Pt: 373.4 °C (760 mm Hg)
Melting Pt: 203…204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06800428
CAS Number: 53590-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR