5-Chloro-1H-indazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A344371-100MG A344371-250MG
103825-904EA 150 USD
103825-904 103836-884 76773-212 76773-214
5-Chloro-1H-indazole-3-carbaldehyde 95%
5-Chloro-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Boiling Pt: 391.7±22.0 °C (760 mm Hg)
Flash Pt: 190.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781535
CAS Number: 102735-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR