4-Chloro-1H-imidazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1H-imidazole

A234385-10G A234385-1G A234385-5G
103810-946EA 27 USD
103810-946 103850-700 76771-222 77089-684 77089-686
4-Chloro-1H-imidazole 98%
4-Chloro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₃ClN₂
MW: 102.52 g/mol
Boiling Pt: 295.9 °C (760 mm Hg)
Melting Pt: 118…120 °C
Flash Pt: 160.8 °C
Storage Temperature: Freezer
MDL Number: MFCD09033087
CAS Number: 15965-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR