5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A120534-10G A120534-250MG A120534-1G
103799-952EA 8 USD
103799-952 76766-428 76766-430 77297-772
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 98%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O
MW: 158.59 g/mol
Boiling Pt: 255.7 °C (760 mm Hg)
Melting Pt: 78…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052538
CAS Number: 27006-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR