5-Chloro-1,2-difluoro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

A196401-100MG A196401-10G A196401-250MG
103816-844EA 64 USD
103816-844 103847-456 76986-300 76986-302 76986-304
5-Chloro-1,2-difluoro-3-nitrobenzene 98%
5-Chloro-1,2-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839707
CAS Number: 169468-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR