5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A139650-250MG A139650-1G A139650-5G
103802-756EA 176 USD
103802-756 77109-756 77315-850 77315-852
5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride 95%
5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N
MW: 204.1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026769
CAS Number: 90562-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR