5-Bromothiophene-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A324110-100G A324110-10G A324110-1G A324110-25G A324110-5G A324110-250MG
103825-362EA 27 USD
103825-362 103836-304 76788-088 76788-090 76839-222 76839-224 76839-226 77668-444
5-Bromothiophene-3-carboxylic acid 97%
5-Bromothiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₂S
Boiling Pt: 318.9 °C (760 mm Hg)
Melting Pt: 140…144 °C
Flash Pt: 146.67 °C
Storage Temperature: Ambient
MDL Number: MFCD08695074
CAS Number: 100523-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR