5-Bromo-2-thenoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

A209845-100G A209845-10G A209845-250MG A209845-500G A209845-1G A209845-25G
103817-960EA 6 USD
103817-960 103848-646 76986-688 76986-690 76986-692 76986-694 77087-002 77087-004
5-Bromo-2-thenoic acid 97%
5-Bromo-2-thenoic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
Melting Pt: 141…144 °C
Flash Pt: 146.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR