5-Bromo-2-thiophenecarbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-thiophenecarbonitrile, 5-Bromothiophene-2-carbonitrile, 2-Cyano-5-bromothiophene, 2-Bromo-5-cyanothiophene
A140821-100G
A140821-250MG
A140821-25G
A140821-1G
A140821-5G
103802-908EA
23
USD
103802-908
76767-768
76767-770
77299-164
77316-022
77316-024
5-Bromo-2-thiophenecarbonitrile 97%
5-Bromo-2-thiophenecarbonitrile
Precaución: for Research Use Only
Formula:
C₅H₂BrNS MW: 188.0500 g/mol Boiling Pt: 226.2 °C (760 mm Hg) Flash Pt: 101.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD04974080 CAS Number: 2160-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...