5-Bromothiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-thiophenecarboxaldehyde

A138385-100G A138385-10G A138385-1G A138385-25G A138385-5G A138385-500G
77162-216EA 90.64 USD
77162-216 77162-218 77162-220 77315-636 77315-638 77592-630
5-Bromothiophene-2-carbaldehyde 98%
5-Bromothiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005432
CAS Number: 4701-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR